N,N-Diisopropyl-2-(5-methoxy-1H-indol-3-yl)-2-oxoacetamide 97%

Supplier: Ambeed
Warning

A831737-100MG A831737-1G A831737-250MG A831737-5G
76653-990EA 16 USD
76653-990 76653-992 76653-994 76653-996
N,N-Diisopropyl-2-(5-methoxy-1H-indol-3-yl)-2-oxoacetamide 97%
N,N-Diisopropyl-2-(5-methoxy-1H-indol-3-yl)-2-oxoacetamide

Precaución: for Research Use Only

Formula: C₁₇H₂₂N₂O₃
MW: 302.3700 g/mol
MDL Number: MFCD16619516
CAS Number: 2426-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR