Difluoromethyl trifluoromethanesulfonate 98%
Supplier: Ambeed
|
Danger
|
Synonyms:
Difluoromethyl triflate, Trifluoromethanesulphonic acid difluoromethyl ester
A831633-1G
A831633-5G
76653-988EA
77.45
USD
76653-988
77749-623
Difluoromethyl trifluoromethanesulfonate 98%
Difluoromethyl trifluoromethanesulfonate
Precaución: for Research Use Only
Formula:
C₂HF₅O₃S MW: 200.085 g/mol |
MDL Number:
MFCD28054469 CAS Number: 1885-46-7 UN: 2920 ADR: 8,3,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...