(4-(Pyrimidin-2-yl)phenyl)methanamine hydrochloride 95%

Supplier: Ambeed
Warning

A831066-100MG A831066-250MG
76653-984EA 198.39 USD
76653-984 76653-986
(4-(Pyrimidin-2-yl)phenyl)methanamine hydrochloride 95%
(4-(Pyrimidin-2-yl)phenyl)methanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₁H₁₂ClN₃
MW: 221.6861 g/mol
MDL Number: MFCD33022484
CAS Number: 2682114-48-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR