Ethyl 2-(3-chloropropoxy)acetate 97%

Supplier: Ambeed
Warning

A830450-1G A830450-5G A830450-25G A830450-10G
76653-980EA 21 USD
76653-980 76653-982 77622-990 77784-171
Ethyl 2-(3-chloropropoxy)acetate 97%
Ethyl 2-(3-chloropropoxy)acetate

Precaución: for Research Use Only

Formula: C₇H₁₃ClO₃
MDL Number: MFCD20621873
CAS Number: 143165-48-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR