[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][bis(1,1-dimethylethyl)[3-methoxy-6-methyl-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine-κP](methanesulfonato-κO)palladium 98%
Supplier: Ambeed
|
Warning
|
A830151-1G
A830151-250MG
A830151-50MG
A830151-5G
76653-972EA
654.58
USD
76653-972
76653-974
76653-976
76653-978
[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][bis(1,1-dimethylethyl)[3-methoxy-6-methyl-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine-κP](methanesulfonato-κO)palladium 98%
[2'-(Amino-κN)[1,1'-biphenyl]-2-yl-κC][bis(1,1-dimethylethyl)[3-methoxy-6-methyl-2',4',6'-tris(1-methylethyl)[1,1'-biphenyl]-2-yl]phosphine-κP](methanesulfonato-κO)palladium
Precaución: for Research Use Only
Formula:
C₄₄H₆₂NO₄PPdS MW: 838.43 g/mol |
MDL Number:
MFCD27952542 CAS Number: 2009020-38-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...