7-Chloro-4,4-dimethyl-3,4-dihydronaphthalen-1(2H)-one 95%

Supplier: Ambeed
Warning

A829695-100MG A829695-1G A829695-250MG
76653-964EA 66 USD
76653-964 76653-966 76653-968
7-Chloro-4,4-dimethyl-3,4-dihydronaphthalen-1(2H)-one 95%
7-Chloro-4,4-dimethyl-3,4-dihydronaphthalen-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₂H₁₃ClO
MDL Number: MFCD11590853
CAS Number: 1082267-76-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR