21-Chloro-3,6,9,12,15-pentaoxahenicosan-1-amine hydrochloride 97%
Supplier: Ambeed
|
Warning
|
A828962-100MG
A828962-250MG
A828962-5G
76653-954EA
51
USD
76653-954
77569-546
77784-169
21-Chloro-3,6,9,12,15-pentaoxahenicosan-1-amine hydrochloride 97%
21-Chloro-3,6,9,12,15-pentaoxahenicosan-1-amine hydrochloride
Precaución: For research use only.
Formula:
C₁₆H₃₅Cl₂NO₅ |
MDL Number:
MFCD32011718 CAS Number: 2241669-16-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...