Potassium [(4-methoxybenzyloxy)methyl]trifluoroborate 98%

Supplier: Ambeed
Warning

Synonyms: trifluoro-[(4-methoxyphenyl)methoxymethyl]boranuide, Potassium, Potassium trifluoro(((4-methoxybenzyl)oxy)methyl)borate

A828863-1G A828863-5G A828863-250MG A828863-25G A828863-100MG
76653-950EA 60 USD
76653-950 76653-952 77622-988 77784-168 77784-167
Potassium [(4-methoxybenzyloxy)methyl]trifluoroborate 98%
Potassium [(4-methoxybenzyloxy)methyl]trifluoroborate

Precaución: for Research Use Only

Formula: C₉H₁₁BF₃O₂·K
MW: 258.087 g/mol
MDL Number: MFCD11505925
CAS Number: 1027642-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR