2-(3-Amino-1H-pyrazol-5-yl)ethanol 95%

Supplier: Ambeed
Warning

A827320-100MG A827320-250MG
76653-930EA 128.37 USD
76653-930 76653-932
2-(3-Amino-1H-pyrazol-5-yl)ethanol 95%
2-(3-Amino-1H-pyrazol-5-yl)ethanol

Precaución: for Research Use Only

Formula: C₅H₉N₃O
MDL Number: MFCD28619875
CAS Number: 1425931-98-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR