1-Butyl-1-methylpiperidinium bis(trifluoromethanesulfonyl)imide 99%

Supplier: Ambeed
Warning

Synonyms: N-butyl-N-methyl-piperidinium bis((trifluoromethyl)sulphonyl)imide

A826938-100G A826938-1G A826938-25G A826938-5G
76653-922EA 541.06 USD
76653-922 76653-924 76653-926 76653-928
1-Butyl-1-methylpiperidinium bis(trifluoromethanesulfonyl)imide 99%
1-Butyl-1-methylpiperidinium bis(trifluoromethanesulfonyl)imide

Precaución: for Research Use Only

Formula: C₁₂H₂₂F₆N₂O₄S₂
MW: 436.43 g/mol
MDL Number: MFCD14156017
CAS Number: 623580-02-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR