(2-Bromo-3-methylphenyl)boronic acid 98%

Supplier: Ambeed
Warning

A826890-1G A826890-250MG A826890-5G
76653-916EA 131 USD
76653-916 76653-918 76653-920
(2-Bromo-3-methylphenyl)boronic acid 98%
(2-Bromo-3-methylphenyl)boronic acid

Precaución: for Research Use Only

Formula: C₇H₈BBrO₂
MDL Number: MFCD30834361
CAS Number: 2304635-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR