Cesium formate hydrate 97%

Supplier: Ambeed
Warning

A826796-100G A826796-25G A826796-5G
76653-910EA 102.91 USD
76653-910 76653-912 76653-914
Cesium formate hydrate 97%
Cesium formate hydrate

Precaución: for Research Use Only

Formula: CsOOCH·xH₂O
MDL Number: MFCD00039103
CAS Number: 1266615-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR