4-Iodophenyl trifluoromethanesulfonate 95%

Supplier: Ambeed
Danger

A826574-100MG A826574-250MG A826574-1G A826574-5G
76653-906EA 11 USD
76653-906 76653-908 77622-982 77622-984
4-Iodophenyl trifluoromethanesulfonate 95%
4-Iodophenyl trifluoromethanesulfonate

Precaución: for Research Use Only

Formula: C₇H₄F₃IO₃S
MW: 352.0695 g/mol
MDL Number: MFCD18379773
CAS Number: 109586-39-2
UN: 3082
ADR: 9 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR