4-Chloro-3-methoxy-5-nitrobenzamide 95%

Supplier: Ambeed
Warning

A826107-100MG A826107-5G A826107-1G
76653-902EA 63.66 USD
76653-902 77573-636 77599-138
4-Chloro-3-methoxy-5-nitrobenzamide 95%
4-Chloro-3-methoxy-5-nitrobenzamide

Precaución: for Research Use Only

Formula: C₈H₇ClN₂O₄
MW: 230.6052 g/mol
MDL Number: MFCD31567784
CAS Number: 2137975-62-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR