6-Cyclopropyl-2-methylnicotinonitrile 95%

Supplier: Ambeed
Warning

A826019-100MG A826019-250MG
76653-894EA 31 USD
76653-894 76653-896
6-Cyclopropyl-2-methylnicotinonitrile 95%
6-Cyclopropyl-2-methylnicotinonitrile

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂
MDL Number: MFCD31414994
CAS Number: 1644447-20-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR