(R)-2-((tert-Butoxycarbonyl)amino)-2-(4-isopropoxyphenyl)acetic acid 95%

Supplier: Ambeed
Warning

A825852-100MG A825852-1G A825852-250MG
76653-888EA 33 USD
76653-888 76653-890 76653-892
(R)-2-((tert-Butoxycarbonyl)amino)-2-(4-isopropoxyphenyl)acetic acid 95%
(R)-2-((tert-Butoxycarbonyl)amino)-2-(4-isopropoxyphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₃NO₅
MW: 309.3575 g/mol
MDL Number: MFCD31696617
CAS Number: 876755-79-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR