4-Methoxycarbonyl-2,6-difluorophenylboronic acid 96%

Supplier: Ambeed
Warning

A823417-1G A823417-250MG
76653-868EA 120 USD
76653-868 76653-870
4-Methoxycarbonyl-2,6-difluorophenylboronic acid 96%
4-Methoxycarbonyl-2,6-difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₇BF₂O₄
MDL Number: MFCD03095142
CAS Number: 2022984-58-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR