Guanidine phosphate 98%

Supplier: Ambeed
Warning

A823390-1000G A823390-100G A823390-25G A823390-500G
76653-856EA 33 USD
76653-856 76653-858 76653-860 77784-150
Guanidine phosphate 98%
Guanidine phosphate

Precaución: for Research Use Only

Formula: CH₈N₃O₄P
MDL Number: MFCD00066148
CAS Number: 5423-22-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR