1-(5-Cyanopyridin-2-yl)cyclopropane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

A822647-250MG A822647-50MG
76653-852EA 546.37 USD
76653-852 76653-854
1-(5-Cyanopyridin-2-yl)cyclopropane-1-carboxylic acid 95%
1-(5-Cyanopyridin-2-yl)cyclopropane-1-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₈N₂O₂
MW: 188.1827 g/mol
MDL Number: MFCD31556341
CAS Number: 2135331-49-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR