8-((tert-Butoxycarbonyl)amino)octyl 4-methylbenzenesulfonate 95%

Supplier: Ambeed
Warning

A822304-100MG A822304-250MG
76653-828EA 105.03 USD
76653-828 76653-830
8-((tert-Butoxycarbonyl)amino)octyl 4-methylbenzenesulfonate 95%
8-((tert-Butoxycarbonyl)amino)octyl 4-methylbenzenesulfonate

Precaución: for Research Use Only

Formula: C₂₀H₃₃NO₅S
MW: 399.5447 g/mol
MDL Number: MFCD30829792
CAS Number: 1472656-81-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR