(2-(Methylsulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 95%

Supplier: Ambeed
Warning

A820258-100MG A820258-1G A820258-250MG
76653-810EA 24 USD
76653-810 76653-812 76653-814
(2-(Methylsulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol 95%
(2-(Methylsulfonyl)-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)methanol

Precaución: for Research Use Only

Formula: C₁₄H₂₁BO₅S
MDL Number: MFCD22494042
CAS Number: 918328-16-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR