6-Bromo-3-fluoroimidazo[1,2-a]pyridine 98%

Supplier: Ambeed
Warning

A819444-100MG A819444-1G A819444-250MG
76653-792EA 68.96 USD
76653-792 76653-794 76653-796
6-Bromo-3-fluoroimidazo[1,2-a]pyridine 98%
6-Bromo-3-fluoroimidazo[1,2-a]pyridine

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.0225 g/mol
MDL Number: MFCD30829494
CAS Number: 2001563-56-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR