2-Amino-N-methylisonicotinamide 95%

Supplier: Ambeed
Warning

A819340-100MG A819340-1G A819340-250MG
76653-786EA 15 USD
76653-786 76653-788 76653-790
2-Amino-N-methylisonicotinamide 95%
2-Amino-N-methylisonicotinamide

Precaución: for Research Use Only

Formula: C₇H₉N₃O
MDL Number: MFCD16688786
CAS Number: 1211295-76-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR