5-Bromo-6-fluoro-2-methyl-1H-indole 95%

Supplier: Ambeed
Warning

A818499-100MG A818499-1G A818499-250MG
76653-780EA 13 USD
76653-780 76653-782 76653-784
5-Bromo-6-fluoro-2-methyl-1H-indole 95%
5-Bromo-6-fluoro-2-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₇BrFN
MW: 228.0610 g/mol
MDL Number: MFCD23706443
CAS Number: 1427363-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR