Tetraethyl orthocarbonate 97%

Supplier: Ambeed
Danger

A817838-25G A817838-5G
76653-776EA 7.43 USD
76653-776 76653-778
Tetraethyl orthocarbonate 97%
Tetraethyl orthocarbonate

Precaución: for Research Use Only

Formula: C₉H₂₀O₄
MW: 192.26 g/mol
MDL Number: MFCD00009221
CAS Number: 78-09-1
UN: 3272
ADR: 3,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR