N'-Butyl-2',3'-difluoro-[1,1'-biphenyl]-4-carbohydrazide 97%

Supplier: Ambeed
Warning

Synonyms: SR-4370

A817824-10MG A817824-1MG A817824-25MG A817824-5MG
76653-768EA 59.42 USD
76653-768 76653-770 76653-772 76653-774
N'-Butyl-2',3'-difluoro-[1,1'-biphenyl]-4-carbohydrazide 97%
N'-Butyl-2',3'-difluoro-[1,1'-biphenyl]-4-carbohydrazide

Precaución: for Research Use Only

Formula: C₁₇H₁₈F₂N₂O
MW: 304.33 g/mol
MDL Number: MFCD31657386
CAS Number: 1816294-67-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR