11-(Bromomethyl)tricosane 97%

Supplier: Ambeed
Warning

A815875-1G A815875-25G A815875-5G
76653-752EA 13.8 USD
76653-752 76653-754 76653-756
11-(Bromomethyl)tricosane 97%
11-(Bromomethyl)tricosane

Precaución: for Research Use Only

Formula: C₂₄H₄₉Br
MW: 417.56 g/mol
CAS Number: 732276-63-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR