1-(tert-Butoxycarbonyl)-5'-methyl-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-2'-carboxylic acid 95%

Supplier: Ambeed
Warning

A815009-100MG A815009-1G A815009-250MG A815009-50MG A815009-5G
76653-734EA 216.43 USD
76653-734 76653-736 76653-738 76653-740 76653-742
1-(tert-Butoxycarbonyl)-5'-methyl-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-2'-carboxylic acid 95%
1-(tert-Butoxycarbonyl)-5'-methyl-4',5'-dihydrospiro[piperidine-4,7'-thieno[2,3-c]pyran]-2'-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₈H₂₅NO₅S
MW: 367.4598 g/mol
MDL Number: MFCD31555888
CAS Number: 2088333-65-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR