TH-34 98%

Supplier: Ambeed
Warning

A813621-10MG A813621-1MG A813621-25MG A813621-50MG A813621-5MG
76653-700EA 40 USD
76653-700 76653-702 76653-704 76653-706 76653-708
TH-34 98%
TH-34

Precaución: for Research Use Only

Formula: C₁₅H₁₆N₂O₂
MDL Number: MFCD31812450
CAS Number: 2196203-96-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR