1-(2-Hydroxyethyl)-3-methylimidazolium chloride 98% (by HPLC)
Supplier: Ambeed
|
Warning
|
A813437-25G
A813437-5G
76653-692EA
37
USD
76653-692
76653-694
1-(2-Hydroxyethyl)-3-methylimidazolium chloride 98% (by HPLC)
1-(2-Hydroxyethyl)-3-methylimidazolium chloride
Precaución: for Research Use Only
Formula:
C₆H₁₁ClN₂O |
MDL Number:
MFCD12761437 CAS Number: 61755-34-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...