(S)-tert-Butyl 2-(cyanomethyl)azetidine-1-carboxylate 95%

Supplier: Ambeed
Warning

A813329-100MG A813329-250MG
76653-688EA 195.21 USD
76653-688 76653-690
(S)-tert-Butyl 2-(cyanomethyl)azetidine-1-carboxylate 95%
(S)-tert-Butyl 2-(cyanomethyl)azetidine-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₆N₂O₂
MW: 196.2462 g/mol
MDL Number: MFCD09751115
CAS Number: 228868-28-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR