3-Formyl-5-(trifluoromethyl)benzonitrile 98%

Supplier: Ambeed
Warning

A812652-1G A812652-250MG
76653-678EA 116 USD
76653-678 76653-680
3-Formyl-5-(trifluoromethyl)benzonitrile 98%
3-Formyl-5-(trifluoromethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₉H₄F₃NO
MDL Number: MFCD22544347
CAS Number: 569370-39-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR