(S)-2-Amino-2-phenylethan-1-ol hydrochloride 95%

Supplier: Ambeed
Warning

A812348-1G A812348-25G A812348-5G
76653-672EA 32 USD
76653-672 76653-674 76653-676
(S)-2-Amino-2-phenylethan-1-ol hydrochloride 95%
(S)-2-Amino-2-phenylethan-1-ol hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₂ClNO
MDL Number: MFCD12910592
CAS Number: 88026-82-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR