(R)-(4-Bromophenyl)(phenyl)methanamine 95%

Supplier: Ambeed
Warning

A812113-100MG A812113-1G
76653-668EA 142.17 USD
76653-668 76653-670
(R)-(4-Bromophenyl)(phenyl)methanamine 95%
(R)-(4-Bromophenyl)(phenyl)methanamine

Precaución: for Research Use Only

Formula: C₁₃H₁₂BrN
MW: 262.1451 g/mol
MDL Number: MFCD22423816
CAS Number: 220441-81-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR