3-Aminobutan-2-one hydrochloride 95%

Supplier: Ambeed
Danger

A811723-100MG A811723-250MG
76653-658EA 89.12 USD
76653-658 76653-660
3-Aminobutan-2-one hydrochloride 95%
3-Aminobutan-2-one hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO
MW: 123.58 g/mol
MDL Number: MFCD16619121
CAS Number: 21419-24-9
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR