2-Chloro-3-(hydroxymethylene)cyclohex-1-enecarbaldehyde 95%

Supplier: Ambeed

A811520-100MG A811520-1G A811520-250MG
76653-652EA 22.28 USD
76653-652 76653-654 76653-656
2-Chloro-3-(hydroxymethylene)cyclohex-1-enecarbaldehyde 95%
2-Chloro-3-(hydroxymethylene)cyclohex-1-enecarbaldehyde

Precaución: for Research Use Only

Formula: C₈H₉ClO₂
MW: 172.6089 g/mol
MDL Number: MFCD01863678
CAS Number: 61010-04-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR