7-Bromo-6-methyl-3,4-dihydronaphthalen-1(2H)-one 95%

Supplier: Ambeed
Warning

A811119-100MG A811119-250MG A811119-50MG
76653-646EA 36 USD
76653-646 76653-648 76653-650
7-Bromo-6-methyl-3,4-dihydronaphthalen-1(2H)-one 95%
7-Bromo-6-methyl-3,4-dihydronaphthalen-1(2H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₁BrO
MDL Number: MFCD18648268
CAS Number: 1273597-18-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR