TP-3654 99%

Supplier: Ambeed
Warning

Synonyms: 2-((1R,4R)-4-((3-(3-(trifluoromethyl)phenyl)imidazo[1,2-b]pyridazin-6-yl)amino)cyclohexyl)propan-2-ol, 2-(trans-4-((3-(3-(Trifluoromethyl)phenyl)imidazo[1,2-b]pyridazin-6-yl)amino)cyclohexyl)propan-2-ol

A809668-10MG A809668-25MG A809668-5MG
76653-622EA 45.62 USD
76653-622 76653-624 76653-626
TP-3654 99%
TP-3654

Precaución: for Research Use Only

Formula: C₂₂H2₂₅F₃N₄O
MW: 418.46 g/mol
MDL Number: MFCD28502206
CAS Number: 1361951-15-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR