7-Chloro-2,3-dihydro-1-benzofuran-3-amine 95%

Supplier: Ambeed
Warning

A809471-100MG A809471-1G A809471-250MG
76653-612EA 46 USD
76653-612 76653-614 76653-616
7-Chloro-2,3-dihydro-1-benzofuran-3-amine 95%
7-Chloro-2,3-dihydro-1-benzofuran-3-amine

Precaución: for Research Use Only

Formula: C₈H₈ClNO
MDL Number: MFCD09026449
CAS Number: 939757-40-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR