Polyethylene glycol 300 ∼ Mn 300
Supplier: Ambeed
|
Warning
|
Synonyms:
PEG 300
A809264-1000G
A809264-100G
A809264-500G
76653-606EA
14
USD
76653-606
76653-608
76653-610
Polyethylene glycol 300 ∼ Mn 300
Polyethylene glycol 300
Precaución: for Research Use Only
Formula:
H(OCH₂CH₂)nOH MW: 300 g/mol Flash Pt: 220 |
MDL Number:
MFCD00081839 CAS Number: 25322-68-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...