2-(3-Chlorophenyl)-1-cyclopropylethan-1-amine 95%

Supplier: Ambeed
Warning

A807764-1G A807764-250MG
76653-600EA 502.87 USD
76653-600 76653-602
2-(3-Chlorophenyl)-1-cyclopropylethan-1-amine 95%
2-(3-Chlorophenyl)-1-cyclopropylethan-1-amine

Precaución: for Research Use Only

Formula: C₁₁H₁₄ClN
MW: 195.6886 g/mol
MDL Number: MFCD11935527
CAS Number: 1181447-41-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR