(1R,2R)-1,2-Bis(2-methoxyphenyl)ethane-1,2-diamine 97%

Supplier: Ambeed
Warning

A807306-100MG A807306-1G A807306-250MG
76653-586EA 22.28 USD
76653-586 76653-588 76653-590
(1R,2R)-1,2-Bis(2-methoxyphenyl)ethane-1,2-diamine 97%
(1R,2R)-1,2-Bis(2-methoxyphenyl)ethane-1,2-diamine

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₂
MW: 272.3422 g/mol
MDL Number: MFCD17013979
CAS Number: 758691-50-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR