(S)-2-(boc-amino)-3-(3-hydroxyphenyl)propionic acid methyl ester 95%

Supplier: Ambeed
Warning

A807186-1G A807186-250MG A807186-5G
76653-576EA 100 USD
76653-576 76653-578 76653-580
(S)-2-(boc-amino)-3-(3-hydroxyphenyl)propionic acid methyl ester 95%
(S)-2-(boc-amino)-3-(3-hydroxyphenyl)propionic acid methyl ester

Precaución: for Research Use Only

Formula: C₁₅H₂₁NO₅
MDL Number: MFCD29917858
CAS Number: 900800-02-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR