(S,S)-1,2-Bis(2-methoxyphenyl)-1,2-ethanediamine 97%

Supplier: Ambeed
Warning

Synonyms: (S,S)-1,2-bis(2-methoxyphenyl)-1,2-ethanediamine

A807174-100MG A807174-1G A807174-250MG
76653-570EA 30.77 USD
76653-570 76653-572 76653-574
(S,S)-1,2-Bis(2-methoxyphenyl)-1,2-ethanediamine 97%
(S,S)-1,2-Bis(2-methoxyphenyl)-1,2-ethanediamine

Precaución: for Research Use Only

Formula: C₁₆H₂₀N₂O₂
MW: 272.348 g/mol
MDL Number: MFCD17013980
CAS Number: 148240-65-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR