4,4'-Dihydroxyazobenzene 98%

Supplier: Ambeed
Warning

Synonyms: 4,4-Dihydroxyazobenzene, 4,4'-Azodiphenol

A804038-100MG A804038-1G A804038-250MG A804038-5G A804038-25G
76653-540EA 7.43 USD
76653-540 76653-542 76653-544 77552-476 77749-591
4,4'-Dihydroxyazobenzene 98%
4,4'-Dihydroxyazobenzene

Precaución: for Research Use Only

Formula: C₁₂H₁₀N₂O₂
MW: 214.22 g/mol
MDL Number: MFCD00045778
CAS Number: 2050-16-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR