(1R)-1-(Thiophen-2-yl)ethan-1-amine 95%

Supplier: Ambeed
Warning

A803890-100MG A803890-1G A803890-250MG A803890-5G
76653-532EA 59.42 USD
76653-532 76653-534 76653-536 76653-538
(1R)-1-(Thiophen-2-yl)ethan-1-amine 95%
(1R)-1-(Thiophen-2-yl)ethan-1-amine

Precaución: for Research Use Only

Formula: C₆H₉NS
MW: 127.2074 g/mol
MDL Number: MFCD09822150
CAS Number: 22038-88-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR