1-Benzofuran-2-yl(pyridin-3-yl)methanamine 2hcl 95%

Supplier: Ambeed
Warning

A803725-100MG A803725-250MG
76653-528EA 83 USD
76653-528 76653-530
1-Benzofuran-2-yl(pyridin-3-yl)methanamine 2hcl 95%
1-Benzofuran-2-yl(pyridin-3-yl)methanamine 2hcl

Precaución: for Research Use Only

Formula: C₁₄H₁₄Cl₂N₂O
MDL Number: MFCD18089486
CAS Number: 1303889-76-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR