3-Bromo-5-fluorobenzylamine hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-5-fluorophenyl)methanamine hydrochloride

A803385-100MG A803385-1G A803385-250MG A803385-5G
76653-518EA 12 USD
76653-518 76653-520 76653-522 77784-097
3-Bromo-5-fluorobenzylamine hydrochloride 98%
3-Bromo-5-fluorobenzylamine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₈BrClFN
MDL Number: MFCD04113608
CAS Number: 1189924-80-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR