(2S,4R)-1-((S)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A802803-100MG A802803-1G A802803-250MG A802803-5G
76653-504EA 11.67 USD
76653-504 76653-506 76653-508 76653-510
(2S,4R)-1-((S)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxylic acid 95%
(2S,4R)-1-((S)-2-((tert-Butoxycarbonyl)amino)-3,3-dimethylbutanoyl)-4-hydroxypyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₈N₂O₆
MW: 344.4033 g/mol
MDL Number: MFCD30729679
CAS Number: 630421-46-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR