2,3,4,6-Tetrachloroanisole 95%

Supplier: Ambeed
Danger

A802551-1G A802551-250MG
76653-484EA 43.5 USD
76653-484 76653-486
2,3,4,6-Tetrachloroanisole 95%
2,3,4,6-Tetrachloroanisole

Precaución: for Research Use Only

Formula: C₇H₄Cl₄O
MW: 245.92 g/mol
MDL Number: MFCD00129714
CAS Number: 938-22-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR